4-(Trifluoromethoxy)benzylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

29770-0010 29770-0050
TS29770-0010EA 42 USD
TS29770-0010 TS29770-0050
4-(Trifluoromethoxy)benzylamine 97%
4-(Trifluoromethoxy)benzylamine
Formula: C₈H₈F₃NO
Boiling Pt: 57…60 °C (10 mmHg)
Density: 1.25 g/cm³
Flash Pt: 77 °C
MDL Number: MFCD00061237
CAS Number: 93919-56-3
EINECS: 300-040-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR