3-Chlorobenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3-Chlorophenyl)methanamine

29757-0050 29757-0250
TS29757-0050EA 53 USD
TS29757-0050 TS29757-0250
3-Chlorobenzylamine 98%
3-Chlorobenzylamine
Formula: C₇H₈ClN
MW: 141.6
Boiling Pt: 110…112 °C (17 mmHg)
Density: 1.159
Flash Pt: 98 °C
MDL Number: MFCD00040752
CAS Number: 4152-90-3
EINECS: 223-985-0
UN: 2735
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5575 to 1.5605 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR