(2-Bromophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

29665-0050 29665-0100
TS29665-0050EA 19 USD
TS29665-0050 TS29665-0100
(2-Bromophenyl)acetic acid 98%
(2-Bromophenyl)acetic acid
Formula: (C₆H₄Br)CH₂COOH
Melting Pt: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00004314
CAS Number: 18698-97-0
EINECS: 242-509-2

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 105 to 110 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR