Terephthalaldehyde mono(diethyl acetal) 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Diethoxymethyl)benzaldehyde

29463-0250 29463-1000
TS29463-0250EA 99 USD
TS29463-0250 TS29463-1000
Terephthalaldehyde mono(diethyl acetal) 97% stabilized
Terephthalaldehyde mono(diethyl acetal)
Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
Boiling Pt: 89…93 °C (7 mmHg)
Density: 1.04 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00010217
CAS Number: 81172-89-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5055 to 1.5085 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer 0.1 % triethylamine

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR