4-Chloro-3-fluorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

29414-0050 29414-0250
TS29414-0050EA 54 USD
TS29414-0050 TS29414-0250
4-Chloro-3-fluorobenzaldehyde 98%
4-Chloro-3-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Melting Pt: 46…49 °C
Flash Pt: 96.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00143288
CAS Number: 5527-95-7

Order Now

Specification Test Results

Appearance White to yellow Crystalline solid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR