5-Bromo-m-xylene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-3,5-dimethylbenzene

29400-0250 29400-1000
TS29400-0250EA 82 USD
TS29400-0250 TS29400-1000
5-Bromo-m-xylene 98%
5-Bromo-m-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Density: 1.36 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000087
CAS Number: 556-96-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5475 to 1.5495 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR