4-Bromo-2-fluorophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

29392-0050 29392-0250 29392-1000
TS29392-0050EA 22 USD
TS29392-0050 TS29392-0250 TS29392-1000
4-Bromo-2-fluorophenol 98%
4-Bromo-2-fluorophenol
Formula: C₆H₄BrFO
Boiling Pt: 79 °C (7 mmHg)
Density: 1.74 g/cm³
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00011722
CAS Number: 2105-94-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5660 to 1.5680 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR