3-Amino-3-phenyl-1-propanol 94%

Supplier: Thermo Scientific Chemicals
Danger

29293-0025 29293-5000
TS29293-0025EA 559 USD
TS29293-0025 TS29293-5000
3-Amino-3-phenyl-1-propanol 94%
3-Amino-3-phenyl-1-propanol
Formula: C₉H₁₃NO
MW: 151.21 g/mol
Melting Pt: 70…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00801124
CAS Number: 14593-04-5
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White Needles
Infrared spectrum Conforms
Melting point 70 to 77 °C
Titration with HClO4 ≥92 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR