Bis(4-chlorophenyl) sulfone 97%

Supplier: Thermo Scientific Chemicals
Warning

29118-0010 29118-2500
TS29118-0010EA 136 USD
TS29118-0010 TS29118-2500
Bis(4-chlorophenyl) sulfone 97%
Bis(4-chlorophenyl)sulphone
Formula: C₁₂H₈Cl₂O₂S
Boiling Pt: 397 °C
Melting Pt: 146…151 °C
Flash Pt: 233 °C
MDL Number: MFCD00000619
CAS Number: 80-07-9
EINECS: 201-247-9

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 146 to 151 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR