(S)-(-)-α,α-Diphenylprolinol 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(-)-α,α-Diphenyl-2-pyrrolidinemethanol

29042-0010 29042-0050
TS29042-0010EA 70 USD
TS29042-0010 TS29042-0050
(S)-(-)-α,α-Diphenylprolinol 99+%
(S)-(-)-α,α-Diphenylprolinol
Formula: C₁₇H₁₉NO
MW: 253.35
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00075506
CAS Number: 112068-01-6

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 77 to 80 °C
GC ≥99.0 %
Specific optical rotation -57° to -61° (25 °C, 589 nm) (c=3, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR