Ethyl-3,4,5-trihydroxybenzoate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Gallic acid ethyl ester, Ethyl 3,4,5-trihydroxybenzoate, Ethyl gallate

27993-1000 27993-5000
TS27993-1000EA 77 USD
TS27993-1000 TS27993-5000
Ethyl-3,4,5-trihydroxybenzoate 99%
Ethyl-3,4,5-trihydroxybenzoate
Formula: C₉H₁₀O₅
MW: 198.18 g/mol
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00016430
CAS Number: 831-61-8
EINECS: 212-608-5

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 151 to 154 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR