2,5-Dibromo-p-xylene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,5-Dibromo-1,4-dimethylbenzene, 1,4-Dibromo-2,5-dimethylbenzene

27901-0500 27901-2500
TS27901-0500EA 81 USD
TS27901-0500 TS27901-2500
2,5-Dibromo-p-xylene 98+%
2,5-Dibromo-p-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 261
Melting Pt: 75
Storage Temperature: Ambient
MDL Number: MFCD00000091
CAS Number: 1074-24-4
EINECS: 214-038-2

Order Now

Specification Test Results

Appearance White to beige to light orange Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 72 to 76 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR