α-Bromo-2-chlorotoulene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(Bromomethyl)-2-chlorobenzene, 2-Chlorobenzyl bromide

27893-0050 27893-0250
TS27893-0050EA 39 USD
TS27893-0050 TS27893-0250
α-Bromo-2-chlorotoulene 97%
α-Bromo-2-chlorotoulene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 103…104 °C (10 mmHg)
Density: 1.586 g/cm³
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00000566
CAS Number: 611-17-6
EINECS: 210-257-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5910 to 1.5930 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR