Diallyl phthalate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Phthalic acid diallyl ester
27662-0010
27662-0025
27662-2500
TS27662-0010EA
92
USD
TS27662-0010
TS27662-0025
TS27662-2500
Diallyl phthalate 98%
Diallyl phthalate
Formula:
C₁₄H₁₄O₄ MW: 246.26 g/mol Boiling Pt: 300 °C (760 mmHg) Melting Pt: –70 °C Density: 1.12 g/cm³ Flash Pt: 166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008646 CAS Number: 131-17-9 EINECS: 205-016-3 UN: 3082 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5180 to 1.5200 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...