(±)-1,2,3,4-Tetrahydro-2-naphthol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,3,4-Tetrahydronaphthalen-2-ol

27188-0010 27188-0050
TS27188-0010EA 177 USD
TS27188-0010 TS27188-0050
(±)-1,2,3,4-Tetrahydro-2-naphthol 97%
(±)-1,2,3,4-Tetrahydro-2-naphthol
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 140 °C (12 mmHg)
Melting Pt: 15.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00045575
CAS Number: 530-91-6
EINECS: 208-497-8

Order Now

Specification Test Results

Appearance Clear light yellow to light brown Viscous liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5670 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR