2,4-Difluorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

27147-0050 27147-0250
TS27147-0050EA 57 USD
TS27147-0050 TS27147-0250
2,4-Difluorobenzaldehyde 98%
2,4-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 65…66 °C (17 mmHg)
Melting Pt: 2…3 °C
Density: 1.29 g/cm³
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00010326
CAS Number: 1550-35-2
EINECS: 216-287-2
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4970 to 1.5000 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR