3,4-Difluorobenzaldehyde 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
27130-0050
27130-0250
TS27130-0050EA
47
USD
TS27130-0050
TS27130-0250
3,4-Difluorobenzaldehyde 96%
3,4-Difluorobenzaldehyde
Formula:
C₇H₄F₂O Boiling Pt: 53 °C (1013 hPa) Density: 1.29 g/cm³ Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010328 CAS Number: 34036-07-2 EINECS: 422-180-3 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4990 to 1.5010 (20°C, 589 nm) |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...