4-Bromo-1,2-dichlorobenzene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-3,4-dichlorobenzene

26981-0250 26981-1000
TS26981-0250EA 77 USD
TS26981-0250 TS26981-1000
4-Bromo-1,2-dichlorobenzene 98+%
4-Bromo-1,2-dichlorobenzene
Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 140
Melting Pt: 24.5
MDL Number: MFCD00040849
CAS Number: 18282-59-2
EINECS: 242-160-6
UN: 2810

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5986 to 1.6006 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR