1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate mixture of isomers

Supplier: Thermo Scientific Chemicals
Warning

26901-0010 26901-2500
TS26901-0010EA 176 USD
TS26901-0010 TS26901-2500
1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate mixture of isomers
1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate
Formula: C₁₀H₇NaO₉S₃·xH₂O
Storage Temperature: Ambient
CAS Number: 19437-42-4

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Loss on drying ≤12 %
HPLC ≥90 % (mix of isomers)
Sodium sulfate (Na2SO4) ≤5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR