3-Fluoropyrocatechol 98+%
Supplier: Thermo Scientific Chemicals
26479-0010
26479-0100
TS26479-0010EA
47
USD
TS26479-0010
TS26479-0100
3-Fluoropyrocatechol 98+%
3-Fluoropyrocatechol
Formula:
C₆H₅FO₂ Boiling Pt: 218 °C (1013 hPa) Melting Pt: 71…74 °C |
MDL Number:
MFCD00042582 CAS Number: 363-52-0 |
Specification Test Results
Appearance | Light beige to grey-brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 69 to 73 °C |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...