1-Chloro-2-iodobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-1-iodobenzene

24946-0050 24946-0500
TS24946-0050EA 30 USD
TS24946-0050 TS24946-0500
1-Chloro-2-iodobenzene 99%
1-Chloro-2-iodobenzene
Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 1 °C
Density: 1.95 g/cm³
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00001033
CAS Number: 615-41-8
EINECS: 210-423-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6330 to 1.6350 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR