1-Bromo-3-fluoro-4-iodobenzene 99+%

Supplier: Thermo Scientific Chemicals
Warning

24940-0250 24940-1000
TS24940-0250EA 58 USD
TS24940-0250 TS24940-1000
1-Bromo-3-fluoro-4-iodobenzene 99+%
1-Bromo-3-fluoro-4-iodobenzene
Formula: C₆H₃BrFI
Melting Pt: 46…49 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00010608
CAS Number: 105931-73-5

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 46 to 49 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR