Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide
24310-0050
24310-0250
TS24310-0010EA
24.37
USD
TS24310-0010
TS24310-0050
TS24310-0250
Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide) 98%
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimide)
Formula:
C₁₉H₁₅NO₅ MW: 337.3 g/mol Melting Pt: 145…153 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00010733 CAS Number: 82911-69-1 EINECS: 433-520-5 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 145 to 153 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...