α,α,α-Trifluoro-2-tolualdehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

23268-0250 23268-1000
TS23268-0250EA 82 USD
TS23268-0250 TS23268-1000
α,α,α-Trifluoro-2-tolualdehyde 97%
α,α,α-Trifluoro-2-tolualdehyde
Formula: C₈H₅F₃O
Boiling Pt: 70…71 °C (16 mmHg)
Melting Pt: –40 °C
Density: 1.32 g/cm³
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00003337
CAS Number: 447-61-0
EINECS: 207-185-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4650 to 1.4670 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR