3,6-Dihydroxyphthalonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-Dicyanohydroquinone, 3,6-Dihydroxyphthalonitrile

23205-0050 23205-0250
TS23205-0050EA 41 USD
TS23205-0050 TS23205-0250
3,6-Dihydroxyphthalonitrile 98%
3,6-Dihydroxyphthalonitrile
Formula: C₈H₄N₂O₂
Melting Pt: >230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001790
CAS Number: 4733-50-0
EINECS: 225-241-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Brown to khaki Crystalline powder or needles
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR