3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 98+%

Supplier: Thermo Scientific Chemicals
Warning

22999-0010 22999-0050
TS22999-0010EA 80 USD
TS22999-0010 TS22999-0050
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 98+%
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate
Formula: C₁₆H₂₀N₂·2HCl·xH₂O
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00150104
CAS Number: 207738-08-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.0 % (on dry substance)
Water ≤12.0 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR