3-Hydroxybenzylhydrazine dihydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

22903-0010 22903-0050
TS22903-0010EA 160 USD
TS22903-0010 TS22903-0050
3-Hydroxybenzylhydrazine dihydrochloride 98%
3-Hydroxybenzylhydrazine dihydrochloride
Formula: C₇H₁₂Cl₂N₂O
MW: 211.086 g/mol
Melting Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00044695
CAS Number: 81012-99-9

Order Now

Specification Test Results

Appearance Off-white to beige Adhering powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR