(3,4-Dihydroxyphenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DOPAC, 3,4-Dihydroxybenzeneacetic acid, 3,4-Dihydroxyphenylacetic acid

22560-0010 22560-0050 22560-0250
TS22560-0010EA 38 USD
TS22560-0010 TS22560-0050 TS22560-0250
(3,4-Dihydroxyphenyl)acetic acid 98%
(3,4-Dihydroxyphenyl)acetic acid
Formula: (C₆H₃(OH)₂)CH₂COOH
MW: 168.15 g/mol
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004338
CAS Number: 102-32-9
EINECS: 203-024-1

Order Now

Specification Test Results

Appearance Beige to light brown to purple-grey Powder
Infrared spectrum Conforms
Melting point 127 to 132 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR