1-Bromo-2-iodobenzene 99% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-1-iodobenzene, 2-Bromoiodobenzene

22195-0050 22195-0250
TS22195-0050EA 49 USD
TS22195-0050 TS22195-0250
1-Bromo-2-iodobenzene 99% stabilized
1-Bromo-2-iodobenzene
Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 120…121 °C (15 mmHg)
Melting Pt: 9…10 °C
Density: 2.203 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001030
CAS Number: 583-55-1
EINECS: 209-508-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6620 to 1.6650 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Copper

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR