1,2-Diiodobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
21840-0050
21840-0200
TS21840-0050EA
109
USD
TS21840-0050
TS21840-0200
1,2-Diiodobenzene 98%
1,2-Diiodobenzene
Formula:
C₆H₄I₂ MW: 329.91 g/mol Boiling Pt: 285 Density: 2.52 g/cm³ Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001038 CAS Number: 615-42-9 EINECS: 210-425-5 |
Specification Test Results
Appearance | Clear yellow to brown Liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...