1-Bromo-4-iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-1-iodobenzene

21838-0250 21838-1000
TS21838-0250EA 120 USD
TS21838-0250 TS21838-1000
1-Bromo-4-iodobenzene 98%
1-Bromo-4-iodobenzene
Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 120…122 °C (14 mmHg)
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00001051
CAS Number: 589-87-7
EINECS: 209-662-7

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 89 to 93 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR