(α,α,α-Trifluoro-4-tolyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-(Trifluoromethyl)phenyl)acetic acid, 4-(Trifluoromethyl)phenylacetic acid

21437-0010 21437-0050
TS21437-0010EA 42 USD
TS21437-0010 TS21437-0050
(α,α,α-Trifluoro-4-tolyl)acetic acid 98%
(α,α,α-Trifluoro-4-tolyl)acetic acid
Formula: (C₆H₄(CF₃))CH₂COOH
MW: 204.15 g/mol
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00004352
CAS Number: 32857-62-8
EINECS: 251-263-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 81 to 85 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR