2,4-Diaminophenol dihydrochloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Hydroxy-1,3-Phenylenediammonium Dichloride, 2,4-Diaminophenol Dihydrochloride, Amidol

21284-0250 21284-1000
TS21284-0250EA 56 USD
TS21284-0250 TS21284-1000
2,4-Diaminophenol dihydrochloride 98%
2,4-Diaminophenol dihydrochloride
Formula: C₆H₁₀Cl₂N₂O
Melting Pt: 222 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012979
CAS Number: 137-09-7
EINECS: 205-279-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige to grey-green Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR