3-Chloro-4-fluoroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
21153-0250
21153-1000
TS21153-0250EA
48
USD
TS21153-0250
TS21153-1000
3-Chloro-4-fluoroaniline 98%
3-Chloro-4-fluoroaniline
Formula:
C₆H₅ClFN MW: 145.56 g/mol Boiling Pt: 230 °C (1013 hPa) Melting Pt: 42…47 °C Flash Pt: 149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007767 CAS Number: 367-21-5 EINECS: 206-682-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Beige to brown Crystalline solid (May darken on storage) |
Infrared spectrum | Conforms |
Melting point | 42 to 47 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...