α,α,α-Trifluoro-4-tolualdehyde 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-(Trifluoromethyl)benzaldehyde, 1-Formyl-4-trifluoromethylbenzene, p-Trifluoromethylbenzaldehyde
20892-0100
20892-0500
TS20892-0100EA
82
USD
TS20892-0100
TS20892-0500
α,α,α-Trifluoro-4-tolualdehyde 98%
α,α,α-Trifluoro-4-tolualdehyde
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4625 to 1.4645 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...