α,α,α-Trifluoro-p-toluidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Aminobenzotrifluoride, 4-(Trifluoromethyl)aniline, Leflunomide impurity A

20891-0100 20891-0500
TS20891-0100EA 59 USD
TS20891-0100 TS20891-0500
α,α,α-Trifluoro-p-toluidine 99%
α,α,α-Trifluoro-p-toluidine
Formula: C₇H₆F₃N
MW: 161.13 g/mol
Boiling Pt: 83 °C (12 mmHg)
Melting Pt: 5…8 °C
Density: 1.296 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00064396
CAS Number: 455-14-1
EINECS: 207-236-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4860 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR