(4-Hydroxyphenyl)acetic acid methyl ester 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-hydroxyphenylacetate, Methyl (4-hydroxyphenyl)acetate, Methyl 2-(4-hydroxyphenyl)acetate

20855-0250 20855-1000
TS20855-0250EA 54 USD
TS20855-0250 TS20855-1000
(4-Hydroxyphenyl)acetic acid methyl ester 99%
(4-Hydroxyphenyl)acetic acid methyl ester
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 162…163 °C (5 mmHg)
Melting Pt: 55…58 °C
MDL Number: MFCD00002387
CAS Number: 14199-15-6
EINECS: 238-050-2

Order Now

Specification Test Results

Appearance White to light yellow Crystals
Infrared spectrum Conforms
Melting point 55 to 58 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR