2,6-Toluenediisocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

20804-0010 20804-0050
TS20804-0010EA 56 USD
TS20804-0010 TS20804-0050
2,6-Toluenediisocyanate 97%
2,6-Toluenediisocyanate
Formula: C₉H₆N₂O₂
Melting Pt: 13 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002010
CAS Number: 91-08-7
EINECS: 202-039-0
UN: 2078
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5700 to 1.5720 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR