Ethyl stearate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Stearic acid ethyl ester, Ethyl octadecanoate

20748-0050 20748-0250
TS20748-0050EA 34 USD
TS20748-0050 TS20748-0250
Ethyl stearate 97%
Ethyl stearate
Formula: C₂₀H₄₀O₂
MW: 312.54 g/mol
Boiling Pt: 365
Melting Pt: 31
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009006
CAS Number: 111-61-5
EINECS: 203-887-4

Order Now

Specification Test Results

Appearance White to light yellow Powder or low melting crystalline mass or liquid
Infrared spectrum Conforms
Melting point 30 to 39 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR