2-Hydroxyiminopropiophenone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Hydroxyiminopropiophenone, 2-Isonitrosopropiophenone, 1-Phenyl-1,2-propanedione-2-oxime, 1-Phenyl-1,2-propanedione 2-Oxime

20502-0100 20502-0250
TS20502-0100EA 36 USD
TS20502-0100 TS20502-0250
2-Hydroxyiminopropiophenone 99%
2-Hydroxyiminopropiophenone
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00002115
CAS Number: 119-51-7
EINECS: 204-329-2

Order Now

Specification Test Results

Appearance White to light yellow Crystals
Infrared spectrum Conforms
Melting point 111 to 115 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR