Pararosaniline base

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(4-aminophenyl)methanol, Paramagenta, p-Rosaniline, Parafuchsin, Pararosaniline base

20155-0100 20155-0500
TS20155-0100EA 67 USD
TS20155-0100 TS20155-0500
Pararosaniline base
Pararosaniline
Formula: C₁₉H₁₉N₃O
MW: 305.38 g/mol
Melting Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00036222
CAS Number: 467-62-9

Order Now

Specification Test Results

Appearance Purple Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR