4-Fluoro-3-nitrobenzotrifluoride 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Fluoro-3-Nitrobenzotrifluoride
20061-0050
20061-1000
TS20061-0050EA
21
USD
TS20061-0050
TS20061-1000
4-Fluoro-3-nitrobenzotrifluoride 96%
4-Fluoro-3-nitrobenzotrifluoride
Formula:
C₇H₃F₄NO₂ MW: 209.1 g/mol Boiling Pt: 92 °C (15 mmHg) Density: 1.49 g/cm³ Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007059 CAS Number: 367-86-2 EINECS: 206-702-5 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4610 to 1.4630 (20°C, 589 nm) |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...