4-Phenoxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals

Synonyms: p-Phenoxybenzenecarbaldehyde, 4-Phenoxybenzenecarbaldehyde

19893-0050 19893-0250
TS19893-0050EA 89 USD
TS19893-0050 TS19893-0250
4-Phenoxybenzaldehyde 98%
4-Phenoxybenzaldehyde
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 185 °C (14 mmHg)
Melting Pt: 24…25 °C
Density: 1.132 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003383
CAS Number: 67-36-7
EINECS: 200-650-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.6100 to 1.6120 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR