4-tert-Butylaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(tert-Butyl)aniline

19825-0050 19825-0250
TS19825-0050EA 61 USD
TS19825-0050 TS19825-0250
4-tert-Butylaniline 99%
4-tert-Butylaniline
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 230 °C
Melting Pt: 15…16 °C
Density: 0.93 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00007899
CAS Number: 769-92-6
EINECS: 212-215-9
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear orange-red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5390 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR