4-Bromobiphenyl (PBB No. 3) 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Bromodiphenyl, PBB No. 3, 4-Bromobiphenyl

19798-0250 19798-1000
TS19798-0250EA 64 USD
TS19798-0250 TS19798-1000
4-Bromobiphenyl (PBB No. 3) 99%
4-Bromobiphenyl (PBB No. 3)
Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 89…92 °C
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00000100
CAS Number: 92-66-0
EINECS: 202-176-6
UN: 3077

Order Now

Specification Test Results

Appearance White to light yellow Crystalline flakes
Infrared spectrum Conforms
Melting point 89 to 92 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR