4-Hexylresorcinol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexylresorcinol, 4-Hexylbenzene-1,3-diol

19792-0250 19792-1000
TS19792-0250EA 70 USD
TS19792-0250 TS19792-1000
4-Hexylresorcinol 98%
4-Hexylresorcinol
Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 65…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00002284
CAS Number: 136-77-6
EINECS: 205-257-4
Merck Index: 12,04750

Order Now

Specification Test Results

Appearance White to light pink to light orange Powder and/or chunks
Infrared spectrum Conforms
Melting point 63 to 68 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR