3-Methoxybenzyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(Chloromethyl)-3-methoxybenzene, 3-(Chloromethyl)anisole
19786-0050
19786-0250
19786-1000
TS19786-0050EA
32
USD
TS19786-0050
TS19786-0250
TS19786-1000
3-Methoxybenzyl chloride 97%
3-Methoxybenzyl chloride
Formula:
C₈H₉ClO Boiling Pt: 124 °C (13 mmHg) Density: 1.07 g/cm³ Flash Pt: 101 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000907 CAS Number: 824-98-6 EINECS: 212-541-1 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5430 to 1.5450 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...