2-Acetyl-1-naphthol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1'-Hydroxy-2'-acetonaphthone, 1-(1-Hydroxynaphthalen-2-yl)ethan-1-one, 2-Acetyl-1-hydroxynaphthalene, 1-Hydroxy-2-acetonaphthone

19500-0050 19500-0250
TS19500-0050EA 22 USD
TS19500-0050 TS19500-0250
2-Acetyl-1-naphthol 99%
2-Acetyl-1-naphthol
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 110…115 °C (0.5 mmHg)
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003963
CAS Number: 711-79-5
EINECS: 211-918-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 97 to 100 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR