(4-Methoxyphenyl)acetic acid ethyl ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl (4-methoxyphenyl)acetate, Ethyl 2-(4-methoxyphenyl)acetate, Homoanisic acid ethyl ester, 4-Methoxyphenylacetic acid ethyl ester, Ethyl 4-Methoxyphenylacetate

18984-0100 189840500
TS18984-0100EA 123 USD
TS18984-0100 TS189840500
(4-Methoxyphenyl)acetic acid ethyl ester 97%
(4-Methoxyphenyl)acetic acid ethyl ester
Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 108…111 °C (3 mmHg)
Density: 1.097 g/cm³
Flash Pt: 46 °C
MDL Number: MFCD00040760
CAS Number: 14062-18-1
EINECS: 237-903-6
UN: 3272

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5065 to 1.5085 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR