4-Methoxyphenylacetone 98+%
Supplier: Thermo Scientific Chemicals
18980-0050
18980-0500
18980-2500
TS18980-0050EA
21
USD
TS18980-0050
TS18980-0500
TS18980-2500
4-Methoxyphenylacetone 98+%
4-Methoxyphenylacetone
Formula:
C₁₀H₁₂O₂ Boiling Pt: 266…268 °C (760 mmHg) Density: 1.067 g/cm³ Flash Pt: 135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008773 CAS Number: 122-84-9 EINECS: 204-578-7 |
Specification Test Results
Appearance | Clear light yellow to yellow-green Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5200 to 1.5300 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...